Anti-KDM2A Goat Polyclonal Antibody
Catalog # EVBIEB10899
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:lysine (K)-specific demethylase 2A
- Antigen symbol:KDM2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Amino acid number:445 to 459
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: KDM2A (lysine (K)-specific demethylase 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: