Anti-MUC5B Goat Polyclonal Antibody
Catalog # EVBIEB10312
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:mucin 5B, oligomeric mucus/gel-forming
- Antigen symbol:MUC5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: MUC5B (mucin 5B, oligomeric mucus/gel-forming)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat