Anti-DSP Goat Polyclonal Antibody
EVBIEB10151
- Antibody type:Primary
- Antigen name:Desmoplakin
- Antigen symbol:DSP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Amino acid number:1672 to 1684
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: DSP (desmoplakin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat