Anti-IGFBP7 Goat Polyclonal Antibody
Catalog # EVBIEB09317
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Insulin-like Growth Factor Binding Protein 7
- Antigen symbol:IGFBP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Amino acid number:145 to 159
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: IGFBP7 (insulin-like growth factor binding protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat