Anti-AMPD1 Goat Polyclonal Antibody
EVBIEB08258
- Antibody type:Primary
- Antigen name:adenosine monophosphate deaminase 1
- Antigen symbol:AMPD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: AMPD1 (adenosine monophosphate deaminase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: