Anti-EPB41L2 Goat polyclonal antibody unconjugated
Catalog # EVBIEB08171
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Erythrocyte membrane protein band 4.1-like 2
- Antigen symbol:EPB41L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Amino acid number:347 to 357
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: EPB41L2 (erythrocyte membrane protein band 4.1-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human