Anti-PTF1A Goat Polyclonal Antibody
EVBIEB07973
- Antibody type:Primary
- Antigen name:Pancreas Specific Transcription Factor 1a
- Antigen symbol:PTF1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Zebrafish
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: PTF1A (pancreas specific transcription factor, 1a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Zebrafish