Anti-PERP Goat Polyclonal Antibody
Catalog # EVBIEB07234
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:PERP, TP53 apoptosis effector
- Antigen symbol:PERP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: PERP (PERP, TP53 apoptosis effector)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat