Anti-DARC Goat polyclonal antibody unconjugated
EVBIEB06940
- Antibody type:Primary
- Antigen name:Duffy blood group, atypical chemokine receptor
- Antigen symbol:DARC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human