Anti-GOLGA6 / GLP Goat polyclonal antibody unconjugated
Catalog # EVBIEB06925
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:GOLGA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:GLP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: GOLGA6 / GLP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human