Anti-HSD11B1 Goat Polyclonal Antibody
EVBIEB06597
- Antibody type:Primary
- Antigen name:hydroxysteroid (11-beta) dehydrogenase 1
- Antigen symbol:HSD11B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: HSD11B1 (hydroxysteroid (11-beta) dehydrogenase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human