Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-E2-EPF / UBE2S Goat Polyclonal Antibody
Catalog # EVBIEB05758
Supplier:  Everest Biotech
Anti-E2-EPF / UBE2S Goat Polyclonal Antibody
Catalog # EVBIEB05758
Supplier:  Everest Biotech

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    E2-EPF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Antigen synonyms:
    UBE2S
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: E2-EPF / UBE2S
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: