Anti-GATA2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1884
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:GATA binding Protein 2
- Antigen symbol:GATA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:MONOMAC|GATA-binding protein 2|NFE1B|DCML|endothelial transcription factor GATA-2
- Amino acid number:46 to 60
- Sequence:DVFFNHLDSQGNPYY
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human GATA2 (46-60aa DVFFNHLDSQGNPYY), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Endothelial transcription factor GATA-2(GATA2) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: GATA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat