Anti-NANOS1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27084
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Nanos homolog 1
- Antigen symbol:NANOS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Gene ID:340719
- Antigen synonyms:EC_Rep1a|NOS-1|NOS1|SPGF12
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NANOS1 (nanos homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: