Anti-IRGC Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-03970
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:IRGC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:56269
- Antigen synonyms:Iigp5|IFGGE|IRGC1|CINEMA|R30953_1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: IRGC (immunity-related GTPase family, cinema)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat