Anti-TLR5 Mouse Monoclonal Antibody [clone: 85B152.5]
Catalog # NOVUNBP1-97728
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 5
- Antigen symbol:TLR5
- Clonality:Monoclonal
- Clone:85B152.5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:7100
- Antigen synonyms:TIL3|Toll-like receptor 5|Toll/interleukin-1 receptor-like protein 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TLR5 (toll-like receptor 5)
Clonality: monoclonal
Clone: 85B152.5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: