Specifications
- Antibody type:Primary
- Antigen name:Very low density lipoprotein receptor
- Antigen symbol:VLDLR
- Clonality:Monoclonal
- Clone:6A6
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:7436
- Antigen synonyms:CAMRQ1|VLDLRCH|CHRMQ1|CARMQ1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: VLDLR (very low density lipoprotein receptor)
Clonality: monoclonal
Clone: 6A6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat