Anti-BNIP3 Rabbit Polyclonal Antibody
The BNIP3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BNIP3. This antibody reacts with human, mouse. The BNIP3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: BNIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- NOVUNBP1-77683
- NOVUNBP1-77683SS
- Antigen symbol:
- BNIP3
- BNIP3
- Antigen name:
- BCL2/adenovirus E1B 19-kDa protein-interacting protein 3
- BCL2/adenovirus E1B 19-kDa protein-interacting protein 3
- Antigen synonyms:
- NIP3
- NIP3
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 664
- 664
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together