Anti-GSK3B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3D10]
Catalog # NOVUNBP1-47470PE
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Glycogen synthase kinase 3 beta
- Antigen symbol:GSK3B
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:
- Environmentally Preferable:
- Gene ID:2932
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: monoclonal
Clone: 3D10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: