Order Entry
Slovenia
ContactUsLinkComponent
Anti-SPP1 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1B20]
Anti-SPP1 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1B20]
  NOVUNB110-89062R
Anti-SPP1 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1B20]
  NOVUNB110-89062R

 

  • Antibody type:
    Primary
  • Antigen name:
    Secreted Phosphoprotein 1
  • Antigen symbol:
    SPP1
  • Clonality:
    Monoclonal
  • Clone:
    1B20
  • Conjugation:
    DyLight® 550
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    6696
  • Antigen synonyms:
    ETA-1|Opnl|Bsp|OP|2AR|BNSP|BSPI|Eta|Ric|Apl-1|Opn|Spp-1|Osteopontin
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: monoclonal
Clone: 1B20
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: