Order Entry
Slovenia
ContactUsLinkComponent
Anti-SOX2 Rabbit Polyclonal Antibody (DyLight® 650)
Anti-SOX2 Rabbit Polyclonal Antibody (DyLight® 650)
  NOVUNB110-37235C
Anti-SOX2 Rabbit Polyclonal Antibody (DyLight® 650)
  NOVUNB110-37235C

 

  • Antibody type:
    Primary
  • Antigen name:
    SRY (sex determining region Y)-box 2
  • Antigen symbol:
    SOX2
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    6657
  • Antigen synonyms:
    MCOPS3|ANOP3|SRY-related HMG-box gene 2|transcription factor SOX-2|transcription factor SOX2|sex determining region Y box 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: