The alpha Tubulin Antibody (DM1A) from Novus Biologicals is a mouse monoclonal antibody to alpha Tubulin. This antibody reacts with human, mouse, rat, porcine, avian, bovine, canine, chicken, chinese hamster, drosophila, fungi, guinea pig, hamster, monkey, parasite, primate, rabbit, xenopus, yeast. The alpha Tubulin Antibody (DM1A) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunomicroscopy, Immunofluorescence, CyTOF-ready.
Simple Western Immunomicroscopy CyTOF-ready
Type: Primary
Antigen: TUBA
Clonality: Monoclonal
Clone: DM1A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Fungi, Guinea Pig, Hamster, Monkey, Parasite, Primate, Rabbit, Xenopus, Yeast
- Catalog No:
- NOVUNB100-690SS
- NOVUNB100-690
- Antigen symbol:
- TUBA1
- TUBA1
- Antigen name:
- Tubulin alpha 1
- Tubulin alpha 1
- Antigen synonyms:
- tubulin alpha-4A chain|alpha 1|tubulin H2-alpha|tubulin|tubulin alpha-1 chain|H2-ALPHA
- tubulin alpha-4A chain|alpha 1|tubulin H2-alpha|tubulin|tubulin alpha-1 chain|H2-ALPHA
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DM1A
- DM1A
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 7846
- 7846
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage buffer:
- PBS
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice