Anti-VEGFA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: VG1]
Catalog # NOVUNB100-664PE
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Vascular endothelial growth factor A
- Antigen symbol:VEGFA
- Clonality:Monoclonal
- Clone:VG1
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:7422
- Antigen synonyms:MVCD1|VEGF|VPF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: VG1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: