Anti-DSG3 Mouse Monoclonal Antibody (Biotin) [clone: 5G11]
NOVUNB100-1643B
- Antibody type:Primary
- Antigen name:Desmoglein 3
- Antigen symbol:DSG3
- Clonality:Monoclonal
- Clone:5G11
- Conjugation:Biotin
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1830
- Antigen synonyms:CDHF6|PVA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5G11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human