Anti-CFC1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB100-1598V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Teratocarcinoma-derived growth factor 1
- Antigen symbol:TDGF1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:6997
- Antigen synonyms:CR|CRGF|CRIPTO
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CFC1 (cripto, FRL-1, cryptic family 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse