Anti-NTHL1 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB100-108B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Endonuclease III like 1
- Antigen symbol:NTH1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:4913
- Antigen synonyms:OCTS 3|Endonuclease III like 1|NTHL 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µL
- Pk:0,2 mL
Specifications
About this item
Type: Primary
Antigen: NTHL1 (nth endonuclease III-like 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat