Order Entry
Slovenia
ContactUsLinkComponent
Anti-Cdk7 Rabbit Polyclonal Antibody
Anti-Cdk7 Rabbit Polyclonal Antibody
  BSBTPA2126-1
undefined
Anti-Cdk7 Rabbit Polyclonal Antibody
  BSBTPA2126-1

 

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent kinase 7
  • Antigen symbol:
    CDK7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    CAK1|MO15|Cell division protein kinase 7|CDK7|CAK|TFIIH basal transcription factor complex kinase subunit|39 kDa protein kinase|P39 Mo15|STK1|CDK-activating kinase
  • Amino acid number:
    329 - 346
  • Sequence:
    RKRTEALEQGGLPKKLIF
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Cdk7 (329-346aa RKRTEALEQGGLPKKLIF), different from the related rat and mouse sequences by two amino acids
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Cyclin-dependent kinase 7(CDK7) detection. Tested with WB in Human, Rat, Mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Cdk7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse