Anti-Caspase-12 Rabbit Polyclonal Antibody
Catalog # BSBTPA2103
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Caspase 12
- Antigen symbol:CASP12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CASP12P1|CASP-12
- Amino acid number:71 - 84
- Sequence:KIFREHLWNSKKQL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Caspase-12 (71-84aa KIFREHLWNSKKQL)
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Inactive caspase-12(CASP12) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Caspase-12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human