Anti-Flavin containing monooxygenase 4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1883
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Flavin containing monooxygenase 4
- Antigen symbol:FMO4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:75 - 92
- Sequence:HEDYPNFMNHEKFWDYLQ
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Flavin containing monooxygenase 4 (75-92aa HEDYPNFMNHEKFWDYLQ), different from the related rat sequence by two amino acids, and from the related mouse sequence by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Dimethylaniline monooxygenase[N-oxide-forming] 4 (FMO4) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Flavin containing monooxygenase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat