Order Entry
Slovenia
ContactUsLinkComponent
Anti-CD163 Rabbit Polyclonal Antibody
Anti-CD163 Rabbit Polyclonal Antibody
Catalog # BSBTPA1874
Supplier:  Boster Bio
undefined
Anti-CD163 Rabbit Polyclonal Antibody
Catalog # BSBTPA1874
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD163 molecule
  • Antigen symbol:
    CD163
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    CD163 antigen|scavenger receptor cysteine-rich type 1 protein M130|hemoglobin scavenger receptor|macrophage-associated antigen
  • Amino acid number:
    1092 - 1109
  • Sequence:
    HQIQYREMNSCLNADDLD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human CD163 (1092-1109aa HQIQYREMNSCLNADDLD).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Scavenger receptor cysteine-rich type 1 protein M130 (CD163) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD163
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human