Anti-cardiac Troponin C Rabbit Polyclonal Antibody
BSBTPA1779
- Antibody type:Primary
- Antigen name:Cardiac Troponin C
- Antigen symbol:TNNC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:TNNC|TNC|CMD1Z|CMH13
- Amino acid number:1 - 18
- Sequence:MDDIYKAAVEQLTEEQKN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human cardiac Troponin C (1-18aa MDDIYKAAVEQLTEEQKN), identical to the related rat and mouse sequences .
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Troponin C, slow skeletal and cardiac muscles (TNNC1) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: cardiac Troponin C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat