Anti-NUR77 Rabbit Polyclonal Antibody
Catalog # BSBTPA1676
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 4 Group A Member 1
- Antigen symbol:NR4A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Orphan nuclear receptor HMR|HMR|ST-59|Orphan nuclear receptor NR4A1|GFRP1|NR4A1|NAK1|Early response protein NAK1|TR3 orphan receptor
- Amino acid number:99 - 112
- Sequence:ASFKFEDFQVYGCY
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human NUR77 (99-112aa ASFKFEDFQVYGCY), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Nuclear receptor subfamily 4 group A member 1 (NR4A1) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: NUR77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat