Order Entry
Slovenia
ContactUsLinkComponent
Anti-Podoplanin/gp36 Rabbit Polyclonal Antibody
Anti-Podoplanin/gp36 Rabbit Polyclonal Antibody
  BSBTPA1674
undefined
Anti-Podoplanin/gp36 Rabbit Polyclonal Antibody
  BSBTPA1674

 

  • Antibody type:
    Primary
  • Antigen name:
    Podoplanin
  • Antigen symbol:
    PDPN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    T1A-2|36-KD|glycoprotein|PA2.26 antigen|hT1alpha-2|hT1alpha-1|AGGRUS|T1A|HT1A-1|GP36|Gp38|PA2.26|CTA-520D8.1|lung type-I cell membrane-associated glycoprotein (T1A-2)|T1-alpha|lung type I cell membrane associated glycoprotein|glycoprotein 36|OTS8|GP40
  • Amino acid number:
    227 - 238
  • Sequence:
    VVMRKMSGRYSP
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Podoplanin/gp36 (227-238aa VVMRKMSGRYSP).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Podoplanin(PDPN) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Podoplanin/gp36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human