Anti-NR4A1 Rabbit Polyclonal Antibody
Catalog # ABCAAB111962-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:nuclear receptor subfamily 4, group A, member 1
- Antigen symbol:NR4A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NR4A1 (nuclear receptor subfamily 4, group A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human