Anti-GTF2B Rabbit Monoclonal Antibody [clone: EP4588]
ABCAAB109518-100
: Abcam
- Antibody type:Primary
- Antigen name:General transcription factor 2B
- Antigen symbol:GTF2B
- Clonality:Monoclonal
- Clone:EP4588
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: GTF2B (general transcription factor IIB)
Clonality: monoclonal
Clone: EP4588
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: