Anti-ITGA2 Rabbit Monoclonal Antibody [clone: EPR5789]
ABCAAB109432-100
: Abcam
- Antibody type:Primary
- Antigen name:Integrin Alpha 2
- Antigen symbol:ITGA2
- Clonality:Monoclonal
- Clone:EPR5789
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: ITGA2 (integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor))
Clonality: monoclonal
Clone: EPR5789
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: