Order Entry
Slovenia
ContactUsLinkComponent
Anti-6X His tag® Mouse Monoclonal Antibody [clone: HIS-1] (AP (Alkaline Phosphatase))
Anti-6X His tag® Mouse Monoclonal Antibody [clone: HIS-1] (AP (Alkaline Phosphatase))
Catalog # ABCAAB49746-100
Supplier:  Abcam
Anti-6X His tag® Mouse Monoclonal Antibody [clone: HIS-1] (AP (Alkaline Phosphatase))
Catalog # ABCAAB49746-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    His Tag
  • Antigen symbol:
    6xHis
  • Clonality:
    Monoclonal
  • Clone:
    HIS-1
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Alkaline Phosphatase Mouse monoclonal [HIS-1] to 6X His tag®.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: 6X His tag®
Clonality: Monoclonal
Clone: HIS-1
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: