Anti-CETP Rabbit polyclonal antibody unconjugated
BIRBORB99181-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Cholesteryl ester transfer protein, plasma
- Antigen symbol:CETP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized
- Amino acid number:368 to 382
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Sequence:PRPDQQHSVAYTFEE
- Storage temperature:Cool
- Concentration:500 ug/ml
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human CETP(368-382aa PRPDQQHSVAYTFEE).
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CETP (cholesteryl ester transfer protein, plasma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human