Order Entry
Slovenia
ContactUsLinkComponent
Anti-HEXIM1 Rabbit Polyclonal Antibody
Anti-HEXIM1 Rabbit Polyclonal Antibody
Catalog # BIRBORB137578-50
Supplier:  Biorbyt
undefined
Anti-HEXIM1 Rabbit Polyclonal Antibody
Catalog # BIRBORB137578-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Hexamethylene bis-acetamide inducible 1
  • Antigen symbol:
    HEXIM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage buffer:
    PBS (without Mg2+, Ca2+), pH 7,4, 150 mM sodium chloride, 0,02% sodium azide, 50% glycerol
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    HEXIM1 antibody was raised against synthetic peptide from human HEXIM1.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

ELISA: 1:10000; Immunofluorescence: 1:100-1:500; Immunochemistry: 20 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: HEXIM1 (hexamethylene bis-acetamide inducible 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat