Anti-MT-ND5 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
BIRBORB129288-100
: Biorbyt
- Antibody type:Primary
- Antigen name:mitochondrially encoded NADH dehydrogenase 5
- Antigen symbol:MT-ND5
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy3®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:67 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human MT-ND5.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: MT-ND5 (mitochondrially encoded NADH dehydrogenase 5)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat