Anti-CEL Mouse monoclonal antibody
Catalog # BIRBORB27493-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Carboxyl ester lipase
- Antigen symbol:CEL
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Format:concentrated
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:N-epsilon-(Carboxyethyl)-Lysine-BSA
- Size:50 µg
- Pk:50 µG
Specifications
About this item
- ELISA: 0.1-0.5 μg/ml; Immunochemistry: 2 μg/ml
Type: Primary
Antigen: CEL (carboxyl ester lipase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: