Anti-Mouse IgG1 IgG2A IgG2B IgG3 Rabbit Polyclonal Antibody
BIRBORB22380-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgG1/IgG2a/IgG2b/IgG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Purified normal IgG, including all known subclasses, isolated from pooled mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: