Anti-IgG2a/IgG2b Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # BIRBORB21388-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG2a/IgG2b
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Goat
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,4, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Pools of purified homogenous IgG2a and IgG2b isolated from rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: Rat IgG2Ab
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: none
Reactivity: