Anti-Bence Jones Lambda Mouse Monoclonal Antibody (Biotin)
Catalog # BIRBORB21183-0.2
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Bence Jones Lambda
- Clonality:Monoclonal
- Conjugation:Biotin
- Host:Mouse
- Reactivity:Human
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:0,4 mg/ml, Biotin/IgG protein molar ratio (B/P) is approximately 2,7, No foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Highly purified Bence Jones lambda proteins isolated from pooled human urine.
- Size:200 μg
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: Bence Jones Lambda
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human