Anti-NCEH1 Goat Polyclonal Antibody
BIRBORB19741-100
: Biorbyt
- Antibody type:Primary
- Antigen name:neutral cholesterol ester hydrolase 1
- Antigen symbol:NCEH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Molecular weight:26 - 28 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C Terminus
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:1000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: NCEH1 (neutral cholesterol ester hydrolase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: