Anti-CPSF7 Mouse Polyclonal Antibody
ABNOH00079869-B01P
: Abnova
- Antibody type:Primary
- Antigen name:cleavage And polyadenylation specific factor 7, 59kDa
- Antigen symbol:CPSF7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79869
- Antigen synonyms:CFIm59
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CPSF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human