Anti-RBFA Mouse Polyclonal Antibody
ABNOH00079863-B02
: Abnova
- Antibody type:Primary
- Antigen name:Ribosome Binding Factor A
- Antigen symbol:RBFA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79863
- Antigen synonyms:HsT169|C18orf22|Chromosome 18 Open Reading Frame 22
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: RBFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human