Anti-PYCRL Rabbit Polyclonal Antibody
Catalog # ABNOH00065263-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pyrroline-5-carboxylate reductase-like
- Antigen symbol:PYCRL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:65263
- Antigen synonyms:PYCR3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PYCRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human