Anti-ZSCAN31 Mouse Monoclonal Antibody [clone: 4F7]
Catalog # ABNOH00064288-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger And SCAN Domain Containing 31
- Antigen symbol:ZSCAN31
- Clonality:Monoclonal
- Clone:4F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:64288
- Antigen synonyms:ZNF20-Lp|ZNF310P|dJ874C20.2|ZNF323
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ZSCAN31
Clonality: Monoclonal
Clone: 4F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human