Anti-MTA3 Mouse Monoclonal Antibody [clone: 2C5]
Catalog # ABNOH00057504-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Metastasis Associated 1 Family Member 3
- Antigen symbol:MTA3
- Clonality:Monoclonal
- Clone:2C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:57504
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MTA3
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human